Search
Search

FBXO9| Gene Symbol: FBXO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012347.4 | NP_036479.1 | 7, 8 | 848 | ||
| NM_033480.2 | NP_258441.1 | 8, 9 | 1098 | |||
| NM_033481.3 | NP_258442.2 | 8, 9 | 975 | |||
| XM_005248995.4 | XP_005249052.1 | 8, 9 | 782 | |||
| XM_005248996.4 | XP_005249053.1 | 8, 9 | 933 | |||
| GenBank mRNA | AB209833.1 | - | 8, 9 | 1137 | ||
| AF174597.1 | - | 4, 5 | 437 | |||
| AF176704.1 | - | 7, 8 | 848 | |||
| AK021559.1 | - | 8, 9 | 1134 | |||
| AK095307.1 | - | 1, 2 | 1651 | |||
| AL137520.1 | - | 8, 9 | 1098 | |||
| AY858798.1 | - | 8, 9 | 1140 | |||
| BC000650.2 | - | 8, 9 | 975 | 3345926 | ||
| BC024973.2 | - | 5, 6 | 519 | |||
| BX641082.1 | - | 1, 2 | 606 | |||