Search
Search

GNG5| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005274.2 | NP_005265.1 | 3 | 503 | ||
| GenBank mRNA | AF038955.1 | - | 2 | 186 | ||
| AF493873.1 | - | 2 | 149 | |||
| AK312111.1 | - | 2 | 262 | |||
| BC003563.1 | - | 2 | 232 | 3502879 | ||
| BM809260.1 | - | 2 | 242 | |||
| BT006823.1 | - | 2 | 149 | |||