Search
Search

SIGLECL1| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173635.2 | NP_775906.1 | 3 | 550 | ||
| XM_005258799.4 | XP_005258856.1 | 3 | 841 | |||
| XM_011526821.2 | XP_011525123.1 | 3 | 463 | |||
| GenBank mRNA | AK097554.1 | - | 3 | 550 | ||
| BC126357.1 | - | 2 | 331 | 8992073 | ||
| BC126359.1 | - | 2 | 331 | 8992075 | ||