Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-E SIRNA Assays ›
          Gene Symbol:
          HLA-E
          Gene Name:
          major histocompatibility complex, class I, E
          Gene Aliases:
          EA1.2, EA2.1, HLA-6.2, MHC, QA1
          Chromosome Location:
          Chr.6: 30489406 - 30494205 on Build GRCh38
          UniGene ID:
          Hs.650174
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS179258
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005516.5 NP_005507.3 6, 7 1151
          XM_017010807.1 XP_016866296.1 5, 6 1163
          XM_017010808.1 XP_016866297.1 5, 6 1163
          GenBank mRNA AB209870.1 - 6, 7 1018
          AJ293263.1 - 6, 7 1025
          AJ293264.1 - 6, 7 1025
          AK098434.1 - 6, 7 1048
          AK225048.1 - 6, 7 1045
          AK292391.1 - 6, 7 1103
          AK296822.1 - 5, 6 1175
          AK298695.1 - 6, 7 871
          AK300906.1 - 6 971
          AK301125.1 - 6, 7 826
          AK314411.1 - 6, 7 1045
          AY216681.1 - 6, 7 1025
          AY221103.1 - 6, 7 1025
          BC002578.2 - 6, 7 1039 3140835
          BC004297.1 - 6, 7 1023
          BC040479.1 - 6, 7 1045 4696327
          BX538084.1 - 5, 6 596
          M20022.1 - 6, 7 1032
          M31183.1 - 6, 7 997
          X56841.1 - 3, 4 312
          X64879.1 - 3, 4 195

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          adaptive immune response
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of endogenous peptide antigen via MHC class Ib
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          regulation of natural killer cell mediated immunity
          positive regulation of natural killer cell mediated immunity
          antibacterial humoral response
          positive regulation of interleukin-13 production
          positive regulation of interleukin-4 production
          positive regulation of TRAIL production
          positive regulation of tumor necrosis factor production
          CD8-positive, alpha-beta T cell activation
          protection from natural killer cell mediated cytotoxicity
          innate immune response
          regulation of immune response
          defense response to Gram-positive bacterium
          positive regulation of immunoglobulin secretion
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          positive regulation of CD8-positive, alpha-beta T cell proliferation
          receptor binding
          beta-2-microglobulin binding
          MHC class I protein binding
          peptide antigen binding
          natural killer cell lectin-like receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0