Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000412.4 | NP_000403.1 | 1 | 184 | ||
XM_005247415.3 | XP_005247472.1 | 1 | 178 | |||
GenBank mRNA | AK313381.1 | - | 1 | 154 | ||
BC069574.1 | - | 1 | 127 | 7262290 | ||
BC150591.1 | - | 1 | 125 | 9056961 | ||
DA637240.1 | - | 1 | 178 |