Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGDIA SIRNA Assays ›
Gene Symbol:
ARHGDIA
Gene Name:
Rho GDP dissociation inhibitor alpha
Gene Aliases:
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
Chromosome Location:
Chr.17: 81867719 - 81871406 on Build GRCh38
UniGene ID:
Hs.159161
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS180674
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGDIA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001185077.2
NP_001172006.1
2
420
NM_001185078.2
NP_001172007.1
2
190
NM_001301240.1
NP_001288169.1
2
190
NM_001301241.1
NP_001288170.1
2
420
NM_001301242.1
NP_001288171.1
2
190
NM_001301243.1
NP_001288172.1
2
190
NM_004309.5
NP_004300.1
2
190
NR_125441.1
2
190
XM_011523574.1
XP_011521876.1
2
415
GenBank mRNA
AF498926.1
-
1
54
AK293147.1
-
2
134
AK300816.1
-
2
163
AK312347.1
-
2
126
BC005851.2
-
2
146
2961556
BC005875.2
-
2
146
2961556
BC008701.3
-
1
81
2966760
BC009759.2
-
2
111
3935956
BC013846.1
-
2
110
BC016031.1
-
1
62
3911525
BC016185.1
-
2
106
4867857
BC024258.1
-
2
106
3027721
BC027730.2
-
2
109
4764249
BC028333.1
-
2
109
BC075827.1
-
2
106
6074977
BC106044.1
-
2
149
6162284
BI755343.1
-
2
111
BI768191.1
-
2
142
BM552638.1
-
2
360
BP325393.1
-
2
121
BQ674863.1
-
2
104
BT006884.1
-
1
54
CF132249.1
-
2
114
CF132324.1
-
2
420
CR456777.1
-
1
54
D13989.1
-
1
74
DA433004.1
-
2
109
DC406556.1
-
2
190
EU794615.1
-
1
81
KU177916.1
-
1
54
KU177917.1
-
1
54
M97579.1
-
1
54
X69550.1
-
2
107
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
negative regulation of cell adhesion
regulation of Rho protein signal transduction
negative regulation of apoptotic process
positive regulation of GTPase activity
negative regulation of axonogenesis
positive regulation of axonogenesis
regulation of small GTPase mediated signal transduction
semaphorin-plexin signaling pathway
regulation of actin cytoskeleton reorganization
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0