Search
Search

SMAD3| Gene Symbol: SMAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145102.1 | NP_001138574.1 | 2 | 423 | ||
| NM_001145103.1 | NP_001138575.1 | 2 | 234 | |||
| NM_005902.3 | NP_005893.1 | 2 | 658 | |||
| XM_011521559.2 | XP_011519861.1 | 2 | 817 | |||
| XM_011521560.1 | XP_011519862.1 | 2 | 597 | |||
| GenBank mRNA | AB209616.1 | - | 2 | 243 | ||
| AK290881.1 | - | 3 | 583 | |||
| AK298139.1 | - | 2 | 234 | |||
| AK316017.1 | - | 2 | 283 | |||
| BC050743.1 | - | 2 | 576 | 6094328 | ||
| DA453132.1 | - | 2 | 423 | |||
| U68019.1 | - | 2 | 426 | |||
| U76622.1 | - | 2 | 397 | |||