Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MBP SIRNA Assays ›
          Gene Symbol:
          MBP
          Gene Name:
          myelin basic protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 76978833 - 77133708 on Build GRCh38
          UniGene ID:
          Hs.551713
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181033
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MBP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025081.1 NP_001020252.1 1 134
          NM_001025090.1 NP_001020261.1 1 134
          NM_001025092.1 NP_001020263.1 1 134
          NM_001025100.1 NP_001020271.1 4 706
          NM_001025101.1 NP_001020272.1 4 706
          NM_002385.2 NP_002376.1 1 134
          XM_011526009.2 XP_011524311.1 4 660
          XM_017025778.1 XP_016881267.1 3 541
          XM_017025780.1 XP_016881269.1 4 660
          GenBank mRNA AB208986.1 - 4 657
          AK098402.1 - 1 90
          AK128770.1 - 1 93
          AK128788.1 - 1 90
          AK289893.1 - 1 89
          AK293922.1 - 1 162
          AK296492.1 - 1 49
          AK314553.1 - 1 130
          BC008749.2 - 1 72 3346521
          BC065248.1 - 4 646 6060520
          BC068550.1 - 1 93
          BC080654.1 - 1 79 4859683
          BC101771.1 - 1 72 8069277
          BC101773.1 - 1 72 8069279
          BC143348.1 - 1 72 9051853
          BC143350.1 - 1 72 9051855
          BG766942.1 - 1 135
          CB153535.1 - 1 92
          CB156561.1 - 1 33
          CR536534.1 - 1 43
          CR541919.1 - 1 43
          CR627018.1 - 1 90
          L18865.1 - 4 657
          M13577.1 - 1 79
          M30047.1 - 1 75
          M30515.1 - 1 50
          M30516.1 - 1 53

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          myelin protein structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          chemical synaptic transmission
          central nervous system development
          aging
          axon ensheathment
          response to toxic substance
          substantia nigra development
          response to progesterone
          response to tumor necrosis factor
          myelination
          response to mercury ion
          negative regulation of axonogenesis
          membrane organization
          response to fatty acid
          protease binding
          protein binding
          structural constituent of myelin sheath

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0