Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB8 SIRNA Assays ›
          Gene Symbol:
          NPIPB8 NPIPB15 NPIPB6 NPIPB9 PDXDC2P
          Gene Name:
          nuclear pore complex interacting protein family member B8
          nuclear pore complex interacting protein family member B15
          nuclear pore complex interacting protein family member B6
          nuclear pore complex interacting protein family member B9
          pyridoxal dependent decarboxylase domain containing 2, pseudogene
          Gene Aliases:
          -
          A-761H5.4, NPIPL2
          NPIPB
          -
          NPIP, PDXDC2
          Chromosome Location:
          Chr.16: 28638065 - 28658693 on Build GRCh38
          Chromosome Location:
          Chr.16: 74374986 - 74392115 on Build GRCh38
          Chromosome Location:
          Chr.16: 28342517 - 28364971 on Build GRCh38
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 69976297 - 70065948 on Build GRCh38
          UniGene ID:
          -
          -
          Hs.726467
          Hs.710214
          Hs.513695
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID HSS181210
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310136.1 NP_001297065.1 7 1204
          XM_017023622.1 XP_016879111.1 8 1475
          XM_017023623.1 XP_016879112.1 9 1412
          XM_017023624.1 XP_016879113.1 8 1328
          XM_017023625.1 XP_016879114.1 8 1308
          XM_017023626.1 XP_016879115.1 9 1947
          Gene Symbol: NPIPB15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306094.1 NP_001293023.1 7 1199
          XM_011523496.2 XP_011521798.1 8 1255
          XM_011523498.2 XP_011521800.1 8 1289
          XM_011523501.2 XP_011521803.1 4 1081
          XM_017023248.1 XP_016878737.1 8 1692
          XM_017023249.1 XP_016878738.1 7 1509
          Gene Symbol: NPIPB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282524.1 NP_001269453.1 7 1221
          XM_005255741.3 XP_005255798.1 8 2033
          XM_005255743.3 XP_005255800.1 4 2612
          XM_011545929.2 XP_011544231.1 9 1853
          XM_011545931.2 XP_011544233.1 8 1486
          XM_017023628.1 XP_016879117.1 8 1236
          XM_017023629.1 XP_016879118.1 9 1737
          XM_017023630.1 XP_016879119.1 9 1506
          XM_017023631.1 XP_016879120.1 7 1850
          XM_017023632.1 XP_016879121.1 9 1194
          GenBank mRNA AI149699.1 - 1 182
          BC144439.1 - 7 1182
          BC157836.1 - 8 1934 9057055
          BQ447892.1 - 1 197
          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287250.1 NP_001274179.1 7 1242
          NM_001287251.1 NP_001274180.1 8 1328
          XM_003118719.7 XP_003118767.2 8 1803
          XM_011545690.2 XP_011543992.1 10 1964
          XM_011545693.2 XP_011543995.1 9 1891
          XM_017022814.1 XP_016878303.1 9 2039
          XM_017022815.1 XP_016878304.1 9 1559
          XM_017022816.1 XP_016878305.1 10 1976
          XM_017022817.1 XP_016878306.1 8 1951
          XM_017022818.1 XP_016878307.1 8 3276
          XM_017022819.1 XP_016878308.1 9 1966
          XM_017022820.1 XP_016878309.1 9 1683
          XM_017022821.1 XP_016878310.1 8 2549
          XM_017022822.1 XP_016878311.1 9 3552
          XM_017022823.1 XP_016878312.1 7 3188
          XM_017022824.1 XP_016878313.1 9 1818
          XM_017022825.1 XP_016878314.1 9 1536
          XM_017022826.1 XP_016878315.1 8 1500
          XM_017022827.1 XP_016878316.1 4 1247
          GenBank mRNA BC171825.1 - 8 1978
          Gene Symbol: PDXDC2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX647358.1 - 2 5847

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0