Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MUTYH SIRNA Assays ›
          Gene Symbol:
          MUTYH
          Gene Name:
          mutY DNA glycosylase
          Gene Aliases:
          MYH
          Chromosome Location:
          Chr.1: 45329242 - 45340925 on Build GRCh38
          UniGene ID:
          Hs.271353
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181380
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MUTYH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001048171.1 NP_001041636.1 11, 12 1159
          NM_001048172.1 NP_001041637.1 11, 12 1050
          NM_001048173.1 NP_001041638.1 11, 12 1047
          NM_001048174.1 NP_001041639.1 11, 12 966
          NM_001128425.1 NP_001121897.1 11, 12 1201
          NM_001293190.1 NP_001280119.1 11, 12 1162
          NM_001293191.1 NP_001280120.1 11, 12 999
          NM_001293192.1 NP_001280121.1 11, 12 1095
          NM_001293195.1 NP_001280124.1 12, 13 1116
          NM_001293196.1 NP_001280125.1 11, 12 984
          NM_012222.2 NP_036354.1 11, 12 1192
          XM_011541497.2 XP_011539799.1 11, 12 1258
          XM_011541498.1 XP_011539800.1 11, 12 1036
          XM_011541499.1 XP_011539801.1 11, 12 1022
          XM_011541500.2 XP_011539802.1 11, 12 1217
          XM_011541501.2 XP_011539803.1 12, 13 1131
          XM_011541502.2 XP_011539804.1 11, 12 1036
          XM_011541503.2 XP_011539805.1 10, 11 1004
          XM_011541504.2 XP_011539806.1 11, 12 1085
          XM_011541505.2 XP_011539807.1 7, 8 705
          XM_011541506.1 XP_011539808.1 7, 8 689
          XM_011541507.2 XP_011539809.2 6, 7 501
          XM_017001331.1 XP_016856820.1 11, 12 1035
          XM_017001332.1 XP_016856821.1 11, 12 1037
          XM_017001333.1 XP_016856822.1 11, 12 1090
          XM_017001334.1 XP_016856823.1 11, 12 983
          XM_017001335.1 XP_016856824.1 11, 12 929
          XM_017001336.1 XP_016856825.1 10, 11 1013
          XM_017001337.1 XP_016856826.1 10, 11 953
          XM_017001338.1 XP_016856827.1 10, 11 1540
          XM_017001339.1 XP_016856828.1 10, 11 991
          GenBank mRNA AB032920.1 - 11, 12 1110
          AB032921.1 - 11, 12 1080
          AB032922.1 - 11, 12 1077
          AB032923.1 - 11, 12 1013
          AB032924.1 - 11, 12 977
          AB032925.1 - 11, 12 944
          AB032926.1 - 12, 13 1094
          AB032927.1 - 11, 12 1015
          AB032928.1 - 11, 12 1012
          AB032929.1 - 11, 12 948
          BC003178.1 - 11, 12 1097 2958002
          KU178203.1 - 11, 12 943

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA glycosylase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          mismatch repair
          response to oxidative stress
          depurination
          DNA binding
          protein binding
          DNA N-glycosylase activity
          MutLalpha complex binding
          MutLbeta complex binding
          MutSalpha complex binding
          MutSbeta complex binding
          metal ion binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-lgjf9:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline