Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NACA SIRNA Assays ›
          Gene Symbol:
          NACA NACAP1
          Gene Name:
          nascent polypeptide-associated complex alpha subunit
          nascent polypeptide associated complex alpha subunit pseudogene 1
          Gene Aliases:
          HSD48, NAC-alpha, NACA1, skNAC
          FKSG17
          Chromosome Location:
          Chr.12: 56712427 - 56726275 on Build GRCh38
          Chromosome Location:
          Chr.8: 101368893 - 101369595 on Build GRCh38
          UniGene ID:
          Hs.505735
          Hs.567608
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS181429
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NACA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113201.2 NP_001106672.1 6 807
          NM_001113202.1 NP_001106673.1 6 730
          NM_001113203.2 NP_001106674.2 9 2860
          NM_001320193.1 NP_001307122.1 6 756
          NM_001320194.1 NP_001307123.1 6 730
          NM_005594.5 NP_005585.1 6 781
          XM_006719412.1 XP_006719475.1 7 7078
          XM_006719413.2 XP_006719476.1 7 6334
          XM_006719414.1 XP_006719477.1 7 6076
          XM_006719418.1 XP_006719481.1 6 513
          XM_006719420.3 XP_006719483.1 6 769
          XM_011538389.1 XP_011536691.1 7 6076
          XM_017019337.1 XP_016874826.1 8 2650
          GenBank mRNA AF054187.1 - 6 757
          AK090650.1 - 6 2387
          AK096699.1 - 9 2617
          AK223085.1 - 6 487
          AK293209.1 - 6 349
          AK311904.1 - 6 476
          AW021139.1 - 2 120
          AY034001.1 - 5 451
          AY605660.1 - 6 486
          AY911673.1 - 6 473
          BC105120.1 - 6 648
          BC105122.1 - 6 648
          BC106041.1 - 6 771
          CD246364.1 - 6 745
          CR450295.1 - 5 448
          X80909.1 - 6 473
          Gene Symbol: NACAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002182.1 1 461
          GenBank mRNA AF315951.1 - 2 523
          AK291701.1 - 3 993
          BC069062.1 - 1 461
          BC069411.1 - 1 554
          BC069542.1 - 1 549
          BC074958.2 - 1 549
          BC074959.2 - 1 549

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0