Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRCAM SIRNA Assays ›
          Gene Symbol:
          NRCAM
          Gene Name:
          neuronal cell adhesion molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 108147626 - 108456442 on Build GRCh38
          UniGene ID:
          Hs.21422
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181574
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRCAM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037132.2 NP_001032209.1 30 3870
          NM_001193582.1 NP_001180511.1 31 3971
          NM_001193583.1 NP_001180512.1 30 3914
          NM_001193584.1 NP_001180513.1 29 3878
          NM_005010.4 NP_005001.3 28 3887
          XM_005250373.2 XP_005250430.1 33 4250
          XM_005250383.2 XP_005250440.1 31 3980
          XM_005250385.2 XP_005250442.1 30 3935
          XM_006716003.1 XP_006716066.1 34 4262
          XM_006716007.1 XP_006716070.1 33 4226
          XM_006716012.1 XP_006716075.1 32 3983
          XM_006716014.1 XP_006716077.1 31 3947
          XM_011516253.1 XP_011514555.1 34 4271
          XM_011516255.1 XP_011514557.1 33 4026
          XM_011516256.1 XP_011514558.1 33 4259
          XM_011516257.1 XP_011514559.1 33 4253
          XM_011516258.1 XP_011514560.1 32 4008
          XM_011516259.1 XP_011514561.1 33 4241
          XM_011516261.1 XP_011514563.1 32 4223
          XM_011516262.1 XP_011514564.1 33 4214
          XM_011516267.1 XP_011514569.1 32 3992
          XM_011516268.2 XP_011514570.1 32 4086
          XM_011516269.1 XP_011514571.1 31 3956
          XM_011516270.1 XP_011514572.1 30 3944
          XM_011516271.1 XP_011514573.1 28 3896
          XM_017012236.1 XP_016867725.1 33 4232
          XM_017012237.1 XP_016867726.1 32 4214
          XM_017012238.1 XP_016867727.1 32 4202
          XM_017012239.1 XP_016867728.1 32 4196
          XM_017012240.1 XP_016867729.1 32 4193
          XM_017012241.1 XP_016867730.1 32 3929
          XM_017012242.1 XP_016867731.1 30 3881
          XM_017012243.1 XP_016867732.1 33 4109
          XM_017012244.1 XP_016867733.1 32 4106
          XM_017012245.1 XP_016867734.1 32 4097
          XM_017012246.1 XP_016867735.1 31 3953
          XM_017012247.1 XP_016867736.1 31 3926
          XM_017012248.1 XP_016867737.1 30 3926
          XM_017012249.1 XP_016867738.1 30 3923
          XM_017012250.1 XP_016867739.1 30 3659
          XM_017012251.1 XP_016867740.1 30 3917
          XM_017012252.1 XP_016867741.1 29 3914
          XM_017012253.1 XP_016867742.1 28 3616
          XM_017012254.1 XP_016867743.1 29 3905
          XM_017012255.1 XP_016867744.1 30 3960
          XM_017012256.1 XP_016867745.1 27 3604
          XM_017012257.1 XP_016867746.1 29 3669
          XM_017012258.1 XP_016867747.1 29 3688
          XM_017012259.1 XP_016867748.1 27 3633
          GenBank mRNA AB002341.1 - 29 3806
          AK092330.1 - 3 801
          AK127035.1 - 1 2041
          AK294195.1 - 4 420
          AY528240.1 - 3 397
          BC114570.1 - 29 3802 40083055
          BC115736.1 - 30 3838 40083062
          BX538010.1 - 28 3832

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          neuron migration
          axonogenesis
          axonal fasciculation
          synapse assembly
          central nervous system development
          protein localization
          single organismal cell-cell adhesion
          neuronal action potential propagation
          regulation of axon extension
          retinal ganglion cell axon guidance
          heterotypic cell-cell adhesion
          clustering of voltage-gated sodium channels
          positive regulation of neuron differentiation
          protein binding
          ankyrin binding
          protein binding involved in heterotypic cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0