Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRCAM SIRNA Assays ›
          Gene Symbol:
          NRCAM
          Gene Name:
          neuronal cell adhesion molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 108147626 - 108456442 on Build GRCh38
          UniGene ID:
          Hs.21422
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181576
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRCAM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037132.2 NP_001032209.1 30 4006
          NM_001193582.1 NP_001180511.1 31 4107
          NM_001193583.1 NP_001180512.1 30 4050
          NM_001193584.1 NP_001180513.1 29 4014
          NM_005010.4 NP_005001.3 28 4023
          XM_005250373.2 XP_005250430.1 33 4386
          XM_005250383.2 XP_005250440.1 31 4116
          XM_005250385.2 XP_005250442.1 30 4071
          XM_006716003.1 XP_006716066.1 34 4398
          XM_006716007.1 XP_006716070.1 33 4362
          XM_006716012.1 XP_006716075.1 32 4119
          XM_006716014.1 XP_006716077.1 31 4083
          XM_011516253.1 XP_011514555.1 34 4407
          XM_011516255.1 XP_011514557.1 33 4162
          XM_011516256.1 XP_011514558.1 33 4395
          XM_011516257.1 XP_011514559.1 33 4389
          XM_011516258.1 XP_011514560.1 32 4144
          XM_011516259.1 XP_011514561.1 33 4377
          XM_011516261.1 XP_011514563.1 32 4359
          XM_011516262.1 XP_011514564.1 33 4350
          XM_011516267.1 XP_011514569.1 32 4128
          XM_011516268.2 XP_011514570.1 32 4222
          XM_011516269.1 XP_011514571.1 31 4092
          XM_011516270.1 XP_011514572.1 30 4080
          XM_011516271.1 XP_011514573.1 28 4032
          XM_017012236.1 XP_016867725.1 33 4368
          XM_017012237.1 XP_016867726.1 32 4350
          XM_017012238.1 XP_016867727.1 32 4338
          XM_017012239.1 XP_016867728.1 32 4332
          XM_017012240.1 XP_016867729.1 32 4329
          XM_017012241.1 XP_016867730.1 32 4065
          XM_017012242.1 XP_016867731.1 30 4017
          XM_017012243.1 XP_016867732.1 33 4245
          XM_017012244.1 XP_016867733.1 32 4242
          XM_017012245.1 XP_016867734.1 32 4233
          XM_017012246.1 XP_016867735.1 31 4089
          XM_017012247.1 XP_016867736.1 31 4062
          XM_017012248.1 XP_016867737.1 30 4062
          XM_017012249.1 XP_016867738.1 30 4059
          XM_017012250.1 XP_016867739.1 30 3795
          XM_017012251.1 XP_016867740.1 30 4053
          XM_017012252.1 XP_016867741.1 29 4050
          XM_017012253.1 XP_016867742.1 28 3752
          XM_017012254.1 XP_016867743.1 29 4041
          XM_017012255.1 XP_016867744.1 30 4096
          XM_017012256.1 XP_016867745.1 27 3740
          XM_017012257.1 XP_016867746.1 29 3805
          XM_017012258.1 XP_016867747.1 29 3824
          XM_017012259.1 XP_016867748.1 27 3769
          GenBank mRNA AB002341.1 - 29 3942
          AJ001057.1 - 30 3885
          AK092330.1 - 3 937
          AK127035.1 - 1 2177
          AK294195.1 - 4 556
          AY528240.1 - 3 533
          BC114570.1 - 29 3938 40083055
          BC115736.1 - 30 3974 40083062
          BX538010.1 - 28 3968

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          neuron migration
          axonogenesis
          axonal fasciculation
          synapse assembly
          central nervous system development
          protein localization
          single organismal cell-cell adhesion
          neuronal action potential propagation
          regulation of axon extension
          retinal ganglion cell axon guidance
          heterotypic cell-cell adhesion
          clustering of voltage-gated sodium channels
          positive regulation of neuron differentiation
          protein binding
          ankyrin binding
          protein binding involved in heterotypic cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0