Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SH3GLB1 SIRNA Assays ›
          Gene Symbol:
          SH3GLB1
          Gene Name:
          SH3 domain containing GRB2 like endophilin B1
          Gene Aliases:
          Bif-1, CGI-61, PPP1R70, dJ612B15.2
          Chromosome Location:
          Chr.1: 86704570 - 86748184 on Build GRCh38
          UniGene ID:
          Hs.136309
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS181861
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SH3GLB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206651.1 NP_001193580.1 3, 4 675
          NM_001206652.1 NP_001193581.1 3, 4 675
          NM_001206653.1 NP_001193582.1 2, 3 533
          NM_016009.4 NP_057093.1 3, 4 675
          XM_006710672.2 XP_006710735.1 3, 4 675
          XM_006710673.2 XP_006710736.1 2, 3 503
          XM_011541538.1 XP_011539840.1 3, 4 675
          XM_017001408.1 XP_016856897.1 2, 3 503
          XM_017001409.1 XP_016856898.1 3, 4 675
          XM_017001410.1 XP_016856899.1 3, 4 675
          GenBank mRNA AB007960.2 - 3, 4 640
          AF151819.1 - 3, 4 370
          AF257318.1 - 3, 4 370
          AF263293.1 - 3, 4 350
          AF350371.1 - 3, 4 426
          AK001954.1 - 3, 4 460
          AK225864.1 - 3, 4 495
          AK303710.1 - 2, 3 486
          BC007455.2 - 3, 4 390 2988769

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001011929, XM_006233433, XM_006233435, XM_006233431, BC079085, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          apoptotic process
          positive regulation of autophagy
          regulation of macroautophagy
          regulation of protein stability
          positive regulation of protein oligomerization
          regulation of cytokinesis
          receptor catabolic process
          cellular response to amino acid starvation
          cellular response to glucose starvation
          autophagic cell death
          protein oligomerization
          membrane fission
          cell-cell adhesion
          positive regulation of membrane tubulation
          protein localization to vacuolar membrane
          positive regulation of protein targeting to mitochondrion
          positive regulation of autophagosome assembly
          protein binding
          lipid binding
          identical protein binding
          protein homodimerization activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0