Search
Search

| Gene Symbol: FAM198B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031700.2 | NP_001026870.2 | 2 | 439 | ||
| NM_001128424.1 | NP_001121896.1 | 2 | 439 | |||
| NM_016613.6 | NP_057697.2 | 2 | 484 | |||
| GenBank mRNA | AK172805.1 | - | 2 | 437 | ||
| AL832589.1 | - | 2 | 310 | |||
| AL834177.1 | - | 1 | 15 | |||
| AY358785.1 | - | 2 | 401 | |||
| BC043193.2 | - | 2 | 439 | 5288748 | ||
| BX489659.1 | - | 2 | 631 | |||
| DA887770.1 | - | 2 | 291 | |||
| Gene Symbol: LOC285505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096792.1 | - | 1 | 589 | ||
| AK129723.1 | - | 3 | 326 | |||