Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKAG2 SIRNA Assays ›
Gene Symbol:
PRKAG2
Gene Name:
protein kinase AMP-activated non-catalytic subunit gamma 2
Gene Aliases:
AAKG, AAKG2, CMH6, H91620p, WPWS
Chromosome Location:
Chr.7: 151556114 - 151877231 on Build GRCh38
UniGene ID:
Hs.647072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182034
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKAG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040633.1
NP_001035723.1
14
1904
NM_001304527.1
NP_001291456.1
12
1508
NM_001304531.1
NP_001291460.1
11
926
NM_016203.3
NP_057287.2
14
2173
NM_024429.1
NP_077747.1
10
998
XM_005250002.3
XP_005250059.1
14
2039
XM_005250004.3
XP_005250061.1
14
1985
XM_005250006.4
XP_005250063.1
12
1738
XM_006716021.2
XP_006716084.1
14
1576
XM_011516282.1
XP_011514584.1
14
1573
XM_011516283.1
XP_011514585.1
14
1576
XM_011516284.1
XP_011514586.1
14
1573
XM_011516285.1
XP_011514587.1
11
1418
XM_011516286.1
XP_011514588.1
10
939
XM_011516287.1
XP_011514589.1
13
1649
XM_017012268.1
XP_016867757.1
14
1982
XM_017012269.1
XP_016867758.1
14
1787
XM_017012270.1
XP_016867759.1
14
1563
XM_017012271.1
XP_016867760.1
14
1982
XM_017012272.1
XP_016867761.1
14
1560
XM_017012273.1
XP_016867762.1
12
1723
XM_017012274.1
XP_016867763.1
11
1415
XM_017012275.1
XP_016867764.1
13
1912
XM_017012276.1
XP_016867765.1
14
2008
XM_017012277.1
XP_016867766.1
10
939
XM_017012278.1
XP_016867767.1
13
1649
XM_017012279.1
XP_016867768.1
13
1915
XM_017012280.1
XP_016867769.1
13
1912
XM_017012281.1
XP_016867770.1
13
1938
XM_017012282.1
XP_016867771.1
12
1358
GenBank mRNA
AB025580.1
-
10
911
AF087875.1
-
10
959
AJ249976.1
-
14
1652
AK001887.1
-
10
998
AK301123.1
-
12
1457
AK314646.1
-
10
960
BC020540.1
-
10
879
BC068598.1
-
14
1873
5259756
BT007127.1
-
10
839
CR933725.1
-
2
161
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
glycogen metabolic process
regulation of glycolytic process
protein phosphorylation
negative regulation of protein kinase activity
fatty acid biosynthetic process
ATP biosynthetic process
carnitine shuttle
cell cycle arrest
positive regulation of peptidyl-threonine phosphorylation
sterol biosynthetic process
macroautophagy
regulation of fatty acid metabolic process
activation of protein kinase activity
intracellular signal transduction
regulation of fatty acid biosynthetic process
positive regulation of protein kinase activity
regulation of fatty acid oxidation
regulation of glucose import
negative regulation of protein serine/threonine kinase activity
regulation of signal transduction by p53 class mediator
AMP-activated protein kinase activity
cAMP-dependent protein kinase inhibitor activity
ATP binding
cAMP-dependent protein kinase regulator activity
phosphorylase kinase regulator activity
AMP binding
protein kinase binding
protein kinase activator activity
ADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0