Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIK3CD SIRNA Assays ›
Gene Symbol:
PIK3CD
Gene Name:
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta
Gene Aliases:
APDS, IMD14, P110DELTA, PI3K, p110D
Chromosome Location:
Chr.1: 9629889 - 9729114 on Build GRCh38
UniGene ID:
Hs.518451
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182306
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIK3CD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005026.3
NP_005017.3
10, 11
1555
XM_006710687.2
XP_006710750.1
10, 11
1615
XM_006710689.2
XP_006710752.1
9, 10
1469
XM_017001476.1
XP_016856965.1
10, 11
1661
XM_017001477.1
XP_016856966.1
11, 12
1598
XM_017001478.1
XP_016856967.1
11, 12
1758
XM_017001479.1
XP_016856968.1
10, 11
1627
XM_017001480.1
XP_016856969.1
9, 10
1469
XM_017001481.1
XP_016856970.1
9, 10
1542
XM_017001482.1
XP_016856971.1
9, 10
1464
XM_017001483.1
XP_016856972.1
9, 10
1367
XM_017001484.1
XP_016856973.1
10, 11
1661
XM_017001485.1
XP_016856974.1
9, 10
1367
GenBank mRNA
AB208835.1
-
5, 6
750
AK303425.1
-
3, 4
417
AK314191.1
-
10, 11
1556
BC132919.1
-
9, 10
1469
40146941
BC132921.1
-
9, 10
1469
40146943
BC144264.1
-
9, 10
1469
9052784
BC150297.1
-
10, 11
1502
8860364
DQ263594.1
-
8, 9
1347
DQ263595.1
-
8, 9
1347
DQ263596.1
-
8, 9
1347
JN190435.1
-
9, 10
1582
U57843.1
-
9, 10
1357
U86453.1
-
10, 11
1542
Y10055.2
-
10, 11
1543
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
natural killer cell differentiation
cytokine production
adaptive immune response
mast cell chemotaxis
respiratory burst involved in defense response
protein phosphorylation
phosphatidylinositol biosynthetic process
inflammatory response
signal transduction
positive regulation of gene expression
T cell chemotaxis
phosphatidylinositol 3-kinase signaling
regulation of phosphatidylinositol 3-kinase signaling
natural killer cell activation
T cell differentiation
positive regulation of cell migration
neutrophil chemotaxis
natural killer cell chemotaxis
B cell chemotaxis
phosphatidylinositol-3-phosphate biosynthetic process
T cell activation
B cell activation
mast cell degranulation
innate immune response
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
T cell receptor signaling pathway
B cell receptor signaling pathway
mast cell differentiation
neutrophil extravasation
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0