Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other POLB SIRNA Assays ›
          Gene Symbol:
          POLB
          Gene Name:
          DNA polymerase beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 42338452 - 42371813 on Build GRCh38
          UniGene ID:
          Hs.654484
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS182455
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: POLB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002690.2 NP_002681.1 7, 8 580
          XM_005273535.3 XP_005273592.1 8, 9 682
          XM_005273536.3 XP_005273593.1 8, 9 682
          XM_005273537.3 XP_005273594.1 7, 8 577
          XM_005273538.2 XP_005273595.1 7, 8 592
          XM_005273539.2 XP_005273596.1 6, 7 487
          XM_005273540.4 XP_005273597.1 3, 4 341
          XM_006716353.2 XP_006716416.1 4, 5 226
          XM_017013583.1 XP_016869072.1 5, 6 484
          XM_017013584.1 XP_016869073.1 4, 5 377
          GenBank mRNA AK223537.1 - 7, 8 579
          AK294025.1 - 3, 4 299
          AK314976.1 - 7, 8 541
          BC100288.1 - 7, 8 544 6151069
          BC106909.1 - 7, 8 506 40031807
          CR536503.1 - 7, 8 410
          CR541802.1 - 7, 8 410
          D29013.1 - 7, 8 523
          JQ664683.1 - 7, 8 410
          JQ927442.1 - 4, 5 226
          L11607.1 - 7, 8 410
          M13140.1 - 7, 8 523

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-directed DNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-dependent DNA replication
          DNA repair
          base-excision repair
          base-excision repair, base-free sugar-phosphate removal
          base-excision repair, gap-filling
          base-excision repair, DNA ligation
          pyrimidine dimer repair
          nucleotide-excision repair, DNA gap filling
          inflammatory response
          cellular response to DNA damage stimulus
          salivary gland morphogenesis
          aging
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to gamma radiation
          somatic hypermutation of immunoglobulin genes
          response to ethanol
          lymph node development
          spleen development
          homeostasis of number of cells
          neuron apoptotic process
          response to hyperoxia
          immunoglobulin heavy chain V-D-J recombination
          DNA biosynthetic process
          damaged DNA binding
          DNA-directed DNA polymerase activity
          DNA-(apurinic or apyrimidinic site) lyase activity
          protein binding
          microtubule binding
          lyase activity
          enzyme binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0