Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBFOX1 SIRNA Assays ›
          Gene Symbol:
          RBFOX1
          Gene Name:
          RNA binding protein, fox-1 homolog 1
          Gene Aliases:
          2BP1, A2BP1, FOX-1, FOX1, HRNBP1
          Chromosome Location:
          Chr.16: 5239749 - 7713343 on Build GRCh38
          UniGene ID:
          Hs.459842
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS182619
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBFOX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142333.1 NP_001135805.1 6, 7 1399
          NM_001142334.1 NP_001135806.1 4, 5 631
          NM_001308117.1 NP_001295046.1 6, 7 736
          NM_018723.3 NP_061193.2 6, 7 1399
          NM_145891.2 NP_665898.1 3, 4 723
          NM_145892.2 NP_665899.1 3, 4 723
          NM_145893.2 NP_665900.1 3, 4 723
          XM_005255386.3 XP_005255443.1 3, 4 649
          XM_005255387.3 XP_005255444.1 3, 4 628
          XM_005255388.4 XP_005255445.1 3, 4 684
          XM_005255390.3 XP_005255447.1 6, 7 1521
          XM_005255391.3 XP_005255448.1 5, 6 531
          XM_005255394.4 XP_005255451.1 3, 4 628
          XM_011522546.2 XP_011520848.1 3, 4 628
          XM_011522547.2 XP_011520849.1 3, 4 628
          XM_011522548.2 XP_011520850.1 3, 4 628
          XM_017023318.1 XP_016878807.1 7, 8 1026
          XM_017023319.1 XP_016878808.1 9, 10 1022
          XM_017023320.1 XP_016878809.1 6, 7 1430
          XM_017023321.1 XP_016878810.1 5, 6 2492
          XM_017023322.1 XP_016878811.1 5, 6 2492
          XM_017023323.1 XP_016878812.1 3, 4 649
          XM_017023324.1 XP_016878813.1 6, 7 1430
          XM_017023325.1 XP_016878814.1 4, 5 1028
          XM_017023326.1 XP_016878815.1 6, 7 1521
          XM_017023327.1 XP_016878816.1 3, 4 649
          XM_017023328.1 XP_016878817.1 6, 7 1430
          XM_017023329.1 XP_016878818.1 6, 7 1430
          XM_017023330.1 XP_016878819.1 5, 6 1969
          XM_017023331.1 XP_016878820.1 3, 4 2435
          XM_017023332.1 XP_016878821.1 3, 4 649
          XM_017023333.1 XP_016878822.1 5, 6 531
          XM_017023334.1 XP_016878823.1 3, 4 649
          XM_017023335.1 XP_016878824.1 6, 7 1521
          XM_017023336.1 XP_016878825.1 5, 6 531
          XM_017023337.1 XP_016878826.1 3, 4 649
          XM_017023338.1 XP_016878827.1 5, 6 531
          XM_017023339.1 XP_016878828.1 3, 4 675
          XM_017023340.1 XP_016878829.1 5, 6 531
          XM_017023341.1 XP_016878830.1 6, 7 1521
          XM_017023342.1 XP_016878831.1 5, 6 531
          XM_017023343.1 XP_016878832.1 3, 4 678
          GenBank mRNA AB208826.1 - 1433
          AF094849.1 - 3, 4 724
          AF107203.1 - 6, 7 1397
          AF109106.1 - 3, 4 724
          AF229057.1 - 3, 4 726
          AF448859.1 - 6, 7 1397
          AK001027.1 - 4, 5 631
          AK293951.1 - 6, 7 736
          AL713700.1 - 5, 6 1246
          BC113691.1 - 3, 4 517 8322743
          BC143816.1 - 3, 4 517
          KJ534926.1 - 3, 4 471
          KJ535000.1 - 3, 4 471
          KJ535001.1 - 3, 4 471

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_021477, NM_183188, AY659954, AB041596, AY659955, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA processing
          nervous system development
          RNA splicing
          RNA transport
          nucleotide binding
          RNA binding
          mRNA binding
          protein binding
          protein C-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0