Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FBLIM1 SIRNA Assays ›
Gene Symbol:
FBLIM1
Gene Name:
filamin binding LIM protein 1
Gene Aliases:
CAL, FBLP-1, FBLP1
Chromosome Location:
Chr.1: 15756171 - 15786590 on Build GRCh38
UniGene ID:
Hs.530101
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182631
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FBLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024215.1
NP_001019386.1
5
797
NM_001024216.1
NP_001019387.1
2
319
NM_017556.2
NP_060026.2
6
889
XM_005245900.1
XP_005245957.1
6
891
XM_005245901.1
XP_005245958.1
6
857
XM_005245902.1
XP_005245959.1
7
1079
XM_005245903.1
XP_005245960.1
7
1100
XM_005245909.3
XP_005245966.1
7
1201
XM_006710704.3
XP_006710767.1
7
1519
XM_006710705.1
XP_006710768.1
7
1111
XM_011541616.2
XP_011539918.1
7
1516
XM_011541617.2
XP_011539919.1
7
1100
XM_011541618.2
XP_011539920.1
7
1100
XM_017001519.1
XP_016857008.1
7
1406
XM_017001520.1
XP_016857009.1
7
1383
XM_017001521.1
XP_016857010.1
7
1409
XM_017001522.1
XP_016857011.1
7
1380
XM_017001523.1
XP_016857012.1
6
756
XM_017001524.1
XP_016857013.1
6
727
XM_017001525.1
XP_016857014.1
7
1123
XM_017001526.1
XP_016857015.1
7
1100
GenBank mRNA
AF459643.1
-
5
797
AK027444.1
-
2
337
AK055259.1
-
6
1230
AL133035.1
-
2
72
AY180161.1
-
4
590
BC019895.1
-
7
1033
5139869
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell shape
single organismal cell-cell adhesion
regulation of integrin activation
cell junction assembly
protein binding
zinc ion binding
filamin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0