Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRMU SIRNA Assays ›
          Gene Symbol:
          TRMU
          Gene Name:
          tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
          Gene Aliases:
          LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1
          Chromosome Location:
          Chr.22: 46335401 - 46357340 on Build GRCh38
          UniGene ID:
          Hs.439524
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS183199
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRMU
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282782.1 NP_001269711.1 2 573
          NM_001282783.1 NP_001269712.1 2 573
          NM_001282784.1 NP_001269713.1 2 573
          NM_001282785.1 NP_001269714.1 2 573
          NM_018006.4 NP_060476.2 2 573
          NR_104240.1 2 573
          NR_104241.1 2 573
          XM_005261678.1 XP_005261735.1 2 260
          XM_005261681.1 XP_005261738.1 2 260
          XM_011530271.2 XP_011528573.1 2 336
          XM_011530272.1 XP_011528574.1 2 260
          XM_011530273.1 XP_011528575.1 2 260
          XM_011530274.2 XP_011528576.1 2 280
          XM_011530275.1 XP_011528577.1 2 216
          GenBank mRNA AB178028.1 - 2 260
          AK001002.1 - 2 260
          AK290712.1 - 2 253
          AK295173.1 - 2 260
          AK303020.1 - 2 312
          AY062123.2 - 2 549
          BC027991.1 - 2 231 5000797
          BC080631.1 - 2 235 6536541
          BX952612.1 - 2 201
          CN365141.1 - 2 222
          CR456445.1 - 2 229
          DQ309998.1 - 2 549
          DQ309999.1 - 2 549
          DQ310000.1 - 2 549

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA wobble position uridine thiolation
          mitochondrial tRNA thio-modification
          tRNA binding
          ATP binding
          sulfurtransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0