Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1217 SIRNA Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
SKT
Chromosome Location:
Chr.10: 23694746 - 24547848 on Build GRCh38
UniGene ID:
Hs.445885
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS183462
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098500.2
NP_001091970.1
12, 13
2880
NM_001282767.1
NP_001269696.1
10, 11
2609
NM_001282768.1
NP_001269697.1
10, 11
2609
NM_001282769.1
NP_001269698.1
6, 7
1692
NM_001282770.1
NP_001269699.1
6, 7
1646
NM_001321681.1
NP_001308610.1
7, 8
1797
NM_019590.4
NP_062536.2
11, 12
2714
XM_005252516.3
XP_005252573.1
6, 7
1620
XM_011519552.2
XP_011517854.2
12, 13
2769
XM_011519555.2
XP_011517857.2
11, 12
2687
XM_011519558.2
XP_011517860.2
12, 13
2771
XM_011519559.2
XP_011517861.2
10, 11
2356
XM_011519562.2
XP_011517864.2
12, 13
2769
XM_011519564.2
XP_011517866.1
10, 11
2336
XM_011519565.2
XP_011517867.2
11, 12
2460
XM_011519566.2
XP_011517868.2
10, 11
2583
XM_017016416.1
XP_016871905.1
12, 13
2795
XM_017016417.1
XP_016871906.1
10, 11
2609
XM_017016418.1
XP_016871907.1
10, 11
3431
XM_017016419.1
XP_016871908.1
9, 10
3326
XM_017016420.1
XP_016871909.1
10, 11
2178
XM_017016421.1
XP_016871910.1
12, 13
2388
XM_017016422.1
XP_016871911.1
9, 10
2073
XM_017016423.1
XP_016871912.1
7, 8
1725
XM_017016424.1
XP_016871913.1
7, 8
1797
XM_017016425.1
XP_016871914.1
12, 13
2771
XM_017016426.1
XP_016871915.1
12, 13
2775
XM_017016427.1
XP_016871916.1
11, 12
2436
XM_017016428.1
XP_016871917.1
9, 10
1957
XM_017016429.1
XP_016871918.1
13, 14
2929
GenBank mRNA
AB033043.2
-
12, 13
2880
AB291616.1
-
9, 10
2136
AK091142.1
-
10, 11
2609
AK096446.1
-
9, 10
1997
AK300422.1
-
6, 7
1631
AL833280.1
-
11, 12
2341
BC018764.1
-
6, 7
1460
BC098577.1
-
6, 7
1600
BC136521.1
-
10, 11
2227
9020511
BC144227.1
-
10, 11
2227
9052747
BX640796.1
-
6, 7
1621
BX648451.1
-
6, 7
1621
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic skeletal system development
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0