Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAVS SIRNA Assays ›
          Gene Symbol:
          MAVS
          Gene Name:
          mitochondrial antiviral signaling protein
          Gene Aliases:
          CARDIF, IPS-1, IPS1, VISA
          Chromosome Location:
          Chr.20: 3846799 - 3876123 on Build GRCh38
          UniGene ID:
          Hs.570362
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS183886
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAVS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206491.1 NP_001193420.1 5 1038
          NM_020746.4 NP_065797.2 6 1213
          NR_037921.1 5 1040
          GenBank mRNA AB033097.1 - 5 1119
          AB097003.1 - 6 1176
          AB232371.1 - 5 1113
          AK023799.1 - 6 1168
          AK291785.1 - 5 997
          AK296897.1 - 5 1005
          AL833736.1 - 1 373
          BC044952.1 - 6 1147 5751684
          BX649084.1 - 5 999
          DQ167126.1 - 5 1041
          DQ174270.1 - 5 1041
          DQ181928.1 - 5 1041
          EF467323.1 - 4 866
          EF467324.1 - 4 868
          KC415005.1 - 5 1041

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          activation of innate immune response
          positive regulation of defense response to virus by host
          signal transduction
          negative regulation of type I interferon production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of protein import into nucleus, translocation
          RIG-I signaling pathway
          defense response to bacterium
          positive regulation of transcription factor import into nucleus
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of viral genome replication
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          defense response to virus
          positive regulation of type I interferon-mediated signaling pathway
          cellular response to exogenous dsRNA
          positive regulation of chemokine (C-C motif) ligand 5 production
          positive regulation of IP-10 production
          regulation of peroxisome organization
          signal transducer activity
          protein binding
          protein kinase binding
          CARD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0