Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPRF SIRNA Assays ›
          Gene Symbol:
          PTPRF
          Gene Name:
          protein tyrosine phosphatase, receptor type F
          Gene Aliases:
          BNAH2, LAR
          Chromosome Location:
          Chr.1: 43522238 - 43623672 on Build GRCh38
          UniGene ID:
          Hs.272062
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS184088
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPRF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005271079.3 XP_005271136.1 18, 19 3497
          XM_005271080.3 XP_005271137.1 17, 18 3485
          XM_005271081.3 XP_005271138.1 16, 17 3458
          XM_005271082.3 XP_005271139.1 15, 16 3152
          XM_006710795.3 XP_006710858.1 19, 20 3475
          XM_006710796.3 XP_006710859.1 17, 18 3470
          XM_006710797.3 XP_006710860.1 18, 19 3497
          XM_006710798.3 XP_006710861.1 17, 18 3470
          XM_006710799.3 XP_006710862.1 17, 18 3191
          XM_006710800.3 XP_006710863.1 16, 17 3164
          XM_006710801.3 XP_006710864.1 17, 18 2918
          XM_011541871.2 XP_011540173.1 19, 20 3509
          XM_011541872.2 XP_011540174.1 19, 20 3495
          XM_011541873.2 XP_011540175.1 19, 20 3429
          XM_017001942.1 XP_016857431.1 16, 17 3126
          XM_017001943.1 XP_016857432.1 16, 17 3179
          XM_017001944.1 XP_016857433.1 16, 17 3164
          XM_017001945.1 XP_016857434.1 15, 16 3152
          XM_017001946.1 XP_016857435.1 16, 17 2612
          XM_017001947.1 XP_016857436.1 15, 16 2585
          XM_017001948.1 XP_016857437.1 15, 16 2611
          XM_017001949.1 XP_016857438.1 16, 17 2576
          XM_017001950.1 XP_016857439.1 14, 15 2581
          GenBank mRNA AB177856.1 - 16, 17 2109
          AB177857.1 - 16, 17 3417
          AB209478.1 - 8, 9 2066
          AK125325.1 - 7, 8 1369
          AK127007.1 - 4, 5 312
          BC048768.1 - 16, 17 3428 5502598
          BX537361.1 - 4, 5 1629
          Y00815.1 - 17, 18 3469

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          transmembrane receptor protein tyrosine phosphatase signaling pathway
          chemical synaptic transmission
          cell migration
          neuron projection regeneration
          peptidyl-tyrosine dephosphorylation
          regulation of axon regeneration
          negative regulation of receptor binding
          protein tyrosine phosphatase activity
          transmembrane receptor protein tyrosine phosphatase activity
          heparin binding
          protein complex binding
          chondroitin sulfate proteoglycan binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline