Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRK SIRNA Assays ›
Gene Symbol:
PTPRK
LOC101928140
Gene Name:
protein tyrosine phosphatase, receptor type K
uncharacterized LOC101928140
Gene Aliases:
R-PTP-kappa
-
Chromosome Location:
Chr.6: 127968779 - 128520847 on Build GRCh38
Chromosome Location:
Chr.6: 128027865 - 128086293 on Build GRCh38
UniGene ID:
Hs.155919
Hs.656779
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
HSS184093
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135648.2
NP_001129120.1
12
2433
NM_001291981.1
NP_001278910.1
12
2433
NM_001291982.1
NP_001278911.1
12
2433
NM_001291983.1
NP_001278912.1
11
2161
NM_001291984.1
NP_001278913.1
12
2433
NM_002844.3
NP_002835.2
12
2433
XM_006715537.2
XP_006715600.1
12
2484
XM_011536014.2
XP_011534316.1
12
2484
XM_011536015.2
XP_011534317.1
12
2484
XM_011536016.2
XP_011534318.1
12
2126
XM_011536017.2
XP_011534319.1
13
2724
XM_011536018.2
XP_011534320.1
13
2397
XM_011536020.2
XP_011534322.1
12
2484
XM_011536021.2
XP_011534323.1
12
2484
XM_017011145.1
XP_016866634.1
13
2448
XM_017011146.1
XP_016866635.1
13
2397
XM_017011147.1
XP_016866636.1
13
2378
XM_017011148.1
XP_016866637.1
13
2375
GenBank mRNA
AB209670.1
-
10
1674
AF533875.1
-
12
2198
AK295028.1
-
12
2358
BC140775.1
-
12
2235
9021643
BC144512.1
-
12
2235
9053039
BC144513.1
-
12
2235
9053040
CR749277.1
-
12
2374
L77886.1
-
12
2132
Z70660.1
-
12
2117
Gene Symbol: LOC101928140
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_125849.1
3
248
GenBank mRNA
BC033773.1
-
3
259
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
cell adhesion
signal transduction
transforming growth factor beta receptor signaling pathway
negative regulation of cell proliferation
negative regulation of keratinocyte proliferation
cell migration
negative regulation of cell migration
neuron projection development
protein localization to cell surface
cellular response to reactive oxygen species
cellular response to UV
peptidyl-tyrosine dephosphorylation
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
focal adhesion assembly
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
beta-catenin binding
protein kinase binding
gamma-catenin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0