Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KMT2C SIRNA Assays ›
          Gene Symbol:
          KMT2C LOC101927429
          Gene Name:
          lysine methyltransferase 2C
          lysine methyltransferase 2C pseudogene
          Gene Aliases:
          HALR, MLL3
          -
          Chromosome Location:
          Chr.7: 152134925 - 152436642 on Build GRCh38
          Chromosome Location:
          Chr.1: 144663837 - 144689892 on Build GRCh38
          UniGene ID:
          Hs.647120
          Hs.568236
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS184145
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KMT2C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_170606.2 NP_733751.2 21 3637
          XM_005250025.4 XP_005250082.1 22 3443
          XM_005250026.3 XP_005250083.1 22 3477
          XM_005250027.4 XP_005250084.1 21 3651
          XM_005250028.4 XP_005250085.1 21 3651
          XM_005250031.4 XP_005250088.1 21 3651
          XM_006716077.3 XP_006716140.1 21 3651
          XM_006716078.3 XP_006716141.1 21 3651
          XM_006716079.3 XP_006716142.1 21 3651
          XM_011516450.2 XP_011514752.1 21 3651
          XM_011516451.2 XP_011514753.1 20 3531
          XM_011516452.2 XP_011514754.1 21 3651
          XM_011516453.2 XP_011514755.1 21 3651
          XM_011516454.2 XP_011514756.1 16 2646
          XM_011516456.2 XP_011514758.1 21 3651
          XM_017012480.1 XP_016867969.1 22 3480
          XM_017012481.1 XP_016867970.1 22 3440
          XM_017012482.1 XP_016867971.1 21 3651
          XM_017012483.1 XP_016867972.1 21 3651
          XM_017012484.1 XP_016867973.1 21 3618
          XM_017012485.1 XP_016867974.1 21 3648
          XM_017012486.1 XP_016867975.1 21 3651
          XM_017012487.1 XP_016867976.1 21 3619
          XM_017012488.1 XP_016867977.1 21 3651
          XM_017012489.1 XP_016867978.1 2 158
          GenBank mRNA AB040939.2 - 10 1753
          AF264750.2 - 7 964
          AK025911.1 - 11 1863
          AK095628.1 - 14 2359
          AY024361.1 - 21 3598
          BC151837.1 - 10 1753
          Gene Symbol: LOC101927429
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_136328.1 7 762
          GenBank mRNA AK310441.1 - 7 765

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 methylation
          DNA binding
          protein binding
          zinc ion binding
          transferase activity, transferring acyl groups
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0