Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAN SIRNA Assays ›
          Gene Symbol:
          RAN
          Gene Name:
          RAN, member RAS oncogene family
          Gene Aliases:
          ARA24, Gsp1, TC4
          Chromosome Location:
          Chr.12: 130871879 - 130877678 on Build GRCh38
          UniGene ID:
          Hs.10842
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS184203
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300796.1 NP_001287725.1 4 496
          NM_001300797.1 NP_001287726.1 4 504
          NM_006325.4 NP_006316.1 5 550
          XM_017019772.1 XP_016875261.1 5 665
          XM_017019773.1 XP_016875262.1 4 402
          GenBank mRNA AB062399.1 - 2 217
          AF052578.1 - 4 431
          AF054183.1 - 5 521
          AF501887.1 - 4 407
          AK300934.1 - 4 425
          AK312466.1 - 5 473
          AK316193.1 - 4 424
          AL357616.1 - 5 460
          BC000852.1 - 5 472
          BC004272.1 - 5 435 3615048
          BC014518.2 - 5 464 3830061
          BC014901.2 - 5 450 3910005
          BC016654.1 - 4 763 4342591
          BC022813.1 - 2 190
          BC051908.2 - 5 475 5504629
          BC072000.1 - 5 447 6644326
          BT007271.1 - 4 407
          CR450347.1 - 4 407
          M31469.1 - 4 407

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal large subunit export from nucleus
          ribosomal small subunit export from nucleus
          protein import into nucleus, translocation
          DNA metabolic process
          tRNA export from nucleus
          protein export from nucleus
          mitotic spindle organization
          mitotic nuclear division
          signal transduction
          small GTPase mediated signal transduction
          spermatid development
          viral process
          hippocampus development
          actin cytoskeleton organization
          androgen receptor signaling pathway
          gene silencing by RNA
          positive regulation of protein binding
          cellular protein complex localization
          pre-miRNA export from nucleus
          positive regulation of transcription, DNA-templated
          cell division
          cellular response to mineralocorticoid stimulus
          intracellular transport of virus
          cell-cell adhesion
          protein localization to nucleolus
          chromatin binding
          transcription coactivator activity
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          protein domain specific binding
          protein complex binding
          poly(A) RNA binding
          dynein intermediate chain binding
          androgen receptor binding
          importin-alpha family protein binding
          pre-miRNA binding
          pre-miRNA transporter activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0