Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAP1GAP SIRNA Assays ›
Gene Symbol:
RAP1GAP
Gene Name:
RAP1 GTPase activating protein
Gene Aliases:
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location:
Chr.1: 21596215 - 21669444 on Build GRCh38
UniGene ID:
Hs.148178
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS184210
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAP1GAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145657.1
NP_001139129.1
18
1556
NM_001145658.1
NP_001139130.1
17
1416
NM_002885.2
NP_002876.2
17
1449
XM_005245955.3
XP_005246012.2
17
1461
XM_006710804.2
XP_006710867.2
17
1461
XM_006710805.2
XP_006710868.2
17
1461
XM_017001965.1
XP_016857454.1
16
1463
XM_017001966.1
XP_016857455.1
16
1463
XM_017001967.1
XP_016857456.1
17
1461
XM_017001968.1
XP_016857457.1
17
1496
XM_017001969.1
XP_016857458.1
16
1463
XM_017001970.1
XP_016857459.1
16
1463
XM_017001971.1
XP_016857460.1
17
1415
XM_017001972.1
XP_016857461.1
17
1461
XM_017001973.1
XP_016857462.1
16
1494
XM_017001974.1
XP_016857463.1
17
1461
XM_017001975.1
XP_016857464.1
16
1463
XM_017001976.1
XP_016857465.1
17
1461
XM_017001977.1
XP_016857466.1
17
1461
XM_017001978.1
XP_016857467.1
18
1610
XM_017001979.1
XP_016857468.1
19
1614
XM_017001980.1
XP_016857469.1
17
1415
XM_017001981.1
XP_016857470.1
19
1590
XM_017001982.1
XP_016857471.1
18
1610
XM_017001983.1
XP_016857472.1
20
1822
XM_017001984.1
XP_016857473.1
19
1590
XM_017001985.1
XP_016857474.1
17
1415
XM_017001986.1
XP_016857475.1
19
1627
XM_017001987.1
XP_016857476.1
18
1556
XM_017001988.1
XP_016857477.1
18
1527
XM_017001989.1
XP_016857478.1
19
1627
XM_017001990.1
XP_016857479.1
18
1512
XM_017001991.1
XP_016857480.1
18
1550
GenBank mRNA
AB007943.3
-
18
1494
BC035030.1
-
14
1205
5261027
BC054490.1
-
17
1356
5767775
M64788.1
-
17
1392
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
negative regulation of neuron differentiation
regulation of small GTPase mediated signal transduction
negative regulation of microvillus assembly
negative regulation of GTP binding
negative regulation of thyroid gland epithelial cell proliferation
cellular response to glial cell derived neurotrophic factor
GTPase activity
GTPase activator activity
protein binding
Ras GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0