Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAP1GAP SIRNA Assays ›
          Gene Symbol:
          RAP1GAP
          Gene Name:
          RAP1 GTPase activating protein
          Gene Aliases:
          RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
          Chromosome Location:
          Chr.1: 21596215 - 21669444 on Build GRCh38
          UniGene ID:
          Hs.148178
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS184211
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAP1GAP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145658.1 NP_001139130.1 18, 19 1659
          NM_002885.2 NP_002876.2 18, 19 1692
          XM_017001972.1 XP_016857461.1 18, 19 1704
          XM_017001973.1 XP_016857462.1 17, 18 1737
          XM_017001975.1 XP_016857464.1 17, 18 1706
          XM_017001976.1 XP_016857465.1 18, 19 1704
          XM_017001977.1 XP_016857466.1 18, 19 1704
          XM_017001980.1 XP_016857469.1 18, 19 1658
          XM_017001985.1 XP_016857474.1 18, 19 1658
          XM_017001986.1 XP_016857475.1 20, 21 1870
          XM_017001987.1 XP_016857476.1 19, 20 1799
          XM_017001988.1 XP_016857477.1 19, 20 1770
          XM_017001989.1 XP_016857478.1 20, 21 1870
          XM_017001990.1 XP_016857479.1 19, 20 1755
          XM_017001991.1 XP_016857480.1 19, 20 1793
          GenBank mRNA BC054490.1 - 18, 19 1599 5767775
          M64788.1 - 18, 19 1635

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          regulation of GTPase activity
          positive regulation of GTPase activity
          negative regulation of neuron differentiation
          regulation of small GTPase mediated signal transduction
          negative regulation of microvillus assembly
          negative regulation of GTP binding
          negative regulation of thyroid gland epithelial cell proliferation
          cellular response to glial cell derived neurotrophic factor
          GTPase activity
          GTPase activator activity
          protein binding
          Ras GTPase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0