Search
Search

RFX5| Gene Symbol: RFX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000449.3 | NP_000440.1 | 11 | 1175 | ||
| NM_001025603.1 | NP_001020774.1 | 11 | 1168 | |||
| XM_005245405.1 | XP_005245462.1 | 11 | 1230 | |||
| XM_005245406.3 | XP_005245463.1 | 11 | 1187 | |||
| XM_011509847.1 | XP_011508149.1 | 11 | 1234 | |||
| XM_011509848.1 | XP_011508150.1 | 11 | 1237 | |||
| XM_011509849.1 | XP_011508151.1 | 11 | 1170 | |||
| XM_011509850.1 | XP_011508152.1 | 11 | 1297 | |||
| XM_017001999.1 | XP_016857488.1 | 11 | 1298 | |||
| XM_017002000.1 | XP_016857489.1 | 11 | 1329 | |||
| GenBank mRNA | AK223526.1 | - | 9 | 967 | ||
| AK302891.1 | - | 10 | 1048 | |||
| AK312289.1 | - | 11 | 1148 | |||
| AL050135.1 | - | 3 | 410 | |||
| BC017471.1 | - | 11 | 1102 | 3909092 | ||
| X85786.1 | - | 10 | 1115 | |||