Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBM26 SIRNA Assays ›
          Gene Symbol:
          RBM26
          Gene Name:
          RNA binding motif protein 26
          Gene Aliases:
          ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
          Chromosome Location:
          Chr.13: 79311827 - 79406265 on Build GRCh38
          UniGene ID:
          Hs.558528
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS184613
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBM26
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286631.1 NP_001273560.1 11, 12 2149
          NM_001286632.1 NP_001273561.1 11, 12 2134
          NM_022118.4 NP_071401.3 11, 12 2134
          XM_005266491.4 XP_005266548.1 11, 12 2216
          XM_005266497.1 XP_005266554.1 11, 12 2216
          XM_006719857.1 XP_006719920.1 11, 12 2216
          XM_011535179.1 XP_011533481.1 11, 12 2216
          XM_011535180.2 XP_011533482.1 11, 12 2216
          XM_011535181.1 XP_011533483.1 11, 12 2216
          XM_011535182.1 XP_011533484.1 11, 12 2201
          XM_011535183.2 XP_011533485.1 11, 12 2216
          XM_011535184.1 XP_011533486.1 11, 12 2186
          XM_011535185.2 XP_011533487.1 11, 12 2186
          XM_011535186.2 XP_011533488.1 11, 12 2178
          XM_011535187.2 XP_011533489.1 11, 12 2186
          XM_011535188.2 XP_011533490.1 11, 12 2178
          XM_011535189.1 XP_011533491.1 11, 12 2216
          XM_011535190.1 XP_011533492.1 11, 12 2216
          XM_011535191.2 XP_011533493.1 11, 12 2216
          XM_011535192.2 XP_011533494.1 11, 12 2178
          XM_011535193.2 XP_011533495.1 11, 12 2171
          XM_011535195.1 XP_011533497.1 11, 12 1914
          XM_017020688.1 XP_016876177.1 11, 12 2216
          XM_017020689.1 XP_016876178.1 11, 12 2201
          XM_017020690.1 XP_016876179.1 11, 12 2208
          XM_017020691.1 XP_016876180.1 11, 12 2208
          XM_017020692.1 XP_016876181.1 11, 12 2178
          XM_017020693.1 XP_016876182.1 11, 12 2216
          XM_017020694.1 XP_016876183.1 11, 12 2201
          XM_017020695.1 XP_016876184.1 11, 12 2201
          XM_017020696.1 XP_016876185.1 11, 12 2208
          XM_017020697.1 XP_016876186.1 11, 12 2208
          XM_017020698.1 XP_016876187.1 11, 12 2171
          XM_017020699.1 XP_016876188.1 11, 12 2193
          XM_017020700.1 XP_016876189.1 11, 12 2178
          XM_017020701.1 XP_016876190.1 11, 12 2216
          XM_017020702.1 XP_016876191.1 11, 12 2216
          XM_017020703.1 XP_016876192.1 11, 12 2186
          GenBank mRNA AK027339.1 - 6, 7 917
          AK027456.1 - 5, 6 783
          AK302871.1 - 11, 12 1743
          AL834343.1 - 6, 7 1047
          AX775885.1 - 5, 6 783
          BC041655.1 - 11, 12 1701 4384399
          BC111697.1 - 11, 12 1695 40038080
          BC111739.1 - 11, 12 1711 40038082
          BX648672.1 - 11, 12 1692

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          negative regulation of phosphatase activity
          nucleotide binding
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0