Search
Search

ADGRL4| Gene Symbol: ADGRL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022159.3 | NP_071442.2 | 11 | 1638 | ||
| GenBank mRNA | AB032017.1 | - | 11 | 1599 | ||
| AF192403.1 | - | 9 | 1299 | |||
| AK129766.1 | - | 2 | 90 | |||
| AK172750.1 | - | 8 | 1117 | |||
| AK292894.1 | - | 11 | 1531 | |||
| AK313738.1 | - | 11 | 1637 | |||
| AY358360.1 | - | 11 | 1505 | |||
Set Membership: |
|