Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHACTR4 SIRNA Assays ›
Gene Symbol:
PHACTR4
Gene Name:
phosphatase and actin regulator 4
Gene Aliases:
PPP1R124
Chromosome Location:
Chr.1: 28369574 - 28500369 on Build GRCh38
UniGene ID:
Hs.225641
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS185957
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHACTR4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048183.1
NP_001041648.1
12
2206
NM_023923.3
NP_076412.3
11
2189
XM_005245970.2
XP_005246027.1
12
2213
XM_017002157.1
XP_016857646.1
12
2213
XM_017002158.1
XP_016857647.1
13
2123
XM_017002159.1
XP_016857648.1
13
2372
XM_017002160.1
XP_016857649.1
11
2189
XM_017002161.1
XP_016857650.1
13
2203
XM_017002162.1
XP_016857651.1
14
2265
XM_017002163.1
XP_016857652.1
12
2818
XM_017002164.1
XP_016857653.1
12
2203
XM_017002165.1
XP_016857654.1
13
2176
XM_017002166.1
XP_016857655.1
13
2203
XM_017002167.1
XP_016857656.1
14
2265
XM_017002168.1
XP_016857657.1
13
2176
XM_017002169.1
XP_016857658.1
13
2106
XM_017002170.1
XP_016857659.1
11
2247
XM_017002171.1
XP_016857660.1
14
2340
XM_017002172.1
XP_016857661.1
11
2024
XM_017002173.1
XP_016857662.1
13
2106
GenBank mRNA
AF130081.1
-
6
833
AK023233.1
-
6
1114
AK025436.1
-
13
2193
BC021247.2
-
11
2168
4577873
BC029266.1
-
12
2065
4422413
BC068508.1
-
14
2195
4825777
CR749449.1
-
12
2185
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
neural tube closure
Rho protein signal transduction
actin cytoskeleton organization
positive regulation of catalytic activity
enteric nervous system development
regulation of cell cycle
closure of optic fissure
negative regulation of integrin-mediated signaling pathway
actin binding
protein phosphatase 1 binding
protein phosphatase type 1 activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0