Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAL1 SIRNA Assays ›
          Gene Symbol:
          TAL1
          Gene Name:
          TAL bHLH transcription factor 1, erythroid differentiation factor
          Gene Aliases:
          SCL, TCL5, bHLHa17, tal-1
          Chromosome Location:
          Chr.1: 47216290 - 47232389 on Build GRCh38
          UniGene ID:
          Hs.705618
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186203
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287347.2 NP_001274276.1 4 744
          NM_001290403.1 NP_001277332.1 4 854
          NM_001290404.1 NP_001277333.1 5 859
          NM_001290405.1 NP_001277334.1 4 727
          NM_001290406.1 NP_001277335.1 2 251
          NM_003189.5 NP_003180.1 5 972
          XM_005271160.4 XP_005271217.1 4 2354
          XM_017002187.1 XP_016857676.1 4 1769
          XM_017002188.1 XP_016857677.1 4 961
          XM_017002189.1 XP_016857678.1 4 764
          XM_017002190.1 XP_016857679.1 4 1460
          XM_017002191.1 XP_016857680.1 3 585
          XM_017002193.1 XP_016857682.1 3 321
          GenBank mRNA M61108.1 - 3 1105
          S53245.1 - 4 744
          X51990.1 - 1 69

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          locomotory behavior
          spinal cord association neuron differentiation
          hemopoiesis
          erythrocyte differentiation
          megakaryocyte differentiation
          platelet formation
          basophil differentiation
          positive regulation of protein complex assembly
          embryonic hemopoiesis
          megakaryocyte development
          regulation of cell proliferation
          erythrocyte maturation
          cell fate commitment
          positive regulation of erythrocyte differentiation
          positive regulation of chromatin assembly or disassembly
          positive regulation of transcription, DNA-templated
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of cell division
          astrocyte fate commitment
          definitive hemopoiesis
          hemangioblast cell differentiation
          hematopoietic stem cell differentiation
          regulation of mast cell differentiation
          regulation of stem cell population maintenance
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          RNA polymerase II transcription factor binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          enzyme binding
          histone deacetylase binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0