Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP73 SIRNA Assays ›
          Gene Symbol:
          TP73
          Gene Name:
          tumor protein p73
          Gene Aliases:
          P73
          Chromosome Location:
          Chr.1: 3652565 - 3736201 on Build GRCh38
          UniGene ID:
          Hs.192132
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186396
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP73
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126240.2 NP_001119712.1 3 689
          NM_001126241.2 NP_001119713.1 3 689
          NM_001126242.2 NP_001119714.1 3 689
          NM_001204184.1 NP_001191113.1 5 712
          NM_001204185.1 NP_001191114.1 5 712
          NM_001204186.1 NP_001191115.1 5 712
          NM_001204187.1 NP_001191116.1 5 712
          NM_001204188.1 NP_001191117.1 5 712
          NM_001204189.1 NP_001191118.1 3 689
          NM_001204190.1 NP_001191119.1 3 689
          NM_001204191.1 NP_001191120.1 3 689
          NM_001204192.1 NP_001191121.1 3 522
          NM_005427.3 NP_005418.1 5 712
          GenBank mRNA AB055065.1 - 3 689
          AB055066.1 - 3 689
          AK302118.1 - 3 510
          AK304177.1 - 3 658
          AK304784.1 - 3 639
          AY040827.1 - 3 689
          AY040828.1 - 3 689
          AY040829.1 - 3 689
          BC117251.1 - 5 712 40125802
          BC117253.1 - 5 712 40125804
          DA446656.1 - 2 412
          Y11416.1 - 5 712

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          P53-like transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          kidney development
          mismatch repair
          transcription from RNA polymerase II promoter
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          cell cycle arrest
          regulation of mitotic cell cycle
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to X-ray
          response to organonitrogen compound
          response to gamma radiation
          regulation of gene expression
          viral process
          mitotic G1 DNA damage checkpoint
          cellular response to UV
          response to drug
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of apoptotic process
          negative regulation of JUN kinase activity
          negative regulation of neuron apoptotic process
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of oligodendrocyte differentiation
          protein tetramerization
          negative regulation of cardiac muscle cell proliferation
          positive regulation of cell cycle arrest
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of signal transduction by p53 class mediator
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          p53 binding
          chromatin binding
          damaged DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein kinase binding
          identical protein binding
          transcription regulatory region DNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0