Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TWIST1 SIRNA Assays ›
          Gene Symbol:
          TWIST1
          Gene Name:
          twist family bHLH transcription factor 1
          Gene Aliases:
          ACS3, BPES2, BPES3, CRS, CRS1, CSO, SCS, TWIST, bHLHa38
          Chromosome Location:
          Chr.7: 19113047 - 19117672 on Build GRCh38
          UniGene ID:
          Hs.644998
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS186975
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TWIST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000474.3 NP_000465.1 1 404
          XM_011515496.1 XP_011513798.1 1 151
          GenBank mRNA BC036704.2 - 1 404 4125830
          X99268.1 - 1 163

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ossification
          osteoblast differentiation
          in utero embryonic development
          neuron migration
          neural tube closure
          aortic valve morphogenesis
          mitral valve morphogenesis
          endocardial cushion morphogenesis
          cardiac neural crest cell migration involved in outflow tract morphogenesis
          transcription, DNA-templated
          muscle organ development
          positive regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          negative regulation of phosphatidylinositol 3-kinase signaling
          regulation of bone mineralization
          positive regulation of fatty acid beta-oxidation
          negative regulation of tumor necrosis factor production
          positive regulation of tumor necrosis factor production
          negative regulation of histone phosphorylation
          negative regulation of histone acetylation
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          negative regulation of peroxisome proliferator activated receptor signaling pathway
          outer ear morphogenesis
          odontogenesis
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          negative regulation of osteoblast differentiation
          positive regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          negative regulation of skeletal muscle tissue development
          embryonic cranial skeleton morphogenesis
          positive regulation of epithelial cell proliferation
          palate development
          cranial suture morphogenesis
          embryonic camera-type eye formation
          eyelid development in camera-type eye
          cellular response to growth factor stimulus
          cellular response to hypoxia
          positive regulation of monocyte chemotactic protein-1 production
          positive regulation of DNA-templated transcription, initiation
          positive regulation of cell motility
          negative regulation of oxidative phosphorylation uncoupler activity
          positive regulation of transcription regulatory region DNA binding
          negative regulation of cellular senescence
          positive regulation of interleukin-6 secretion
          negative regulation of double-strand break repair
          cell proliferation involved in heart valve development
          positive regulation of endocardial cushion to mesenchymal transition involved in heart valve formation
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein domain specific binding
          protein homodimerization activity
          bHLH transcription factor binding
          protein heterodimerization activity
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0