Search
Search

ZNF35| Gene Symbol: ZNF35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003420.3 | NP_003411.3 | 4 | 1598 | ||
| XM_017007129.1 | XP_016862618.1 | 4 | 1572 | |||
| XM_017007130.1 | XP_016862619.1 | 4 | 1593 | |||
| XM_017007131.1 | XP_016862620.1 | 4 | 1594 | |||
| GenBank mRNA | AK300581.1 | - | 3 | 1452 | ||
| AK314821.1 | - | 4 | 1597 | |||
| BC013597.1 | - | 4 | 1576 | 4186299 | ||
| BC015892.1 | - | 3 | 1430 | |||
| BT006962.1 | - | 3 | 1353 | |||
| X07289.1 | - | 5 | 1482 | |||