Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRMT2B SIRNA Assays ›
          Gene Symbol:
          TRMT2B
          Gene Name:
          tRNA methyltransferase 2 homolog B
          Gene Aliases:
          CXorf34, dJ341D10.3
          Chromosome Location:
          Chr.X: 100973358 - 101052118 on Build GRCh38
          UniGene ID:
          Hs.496501
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS188304
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRMT2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167970.1 NP_001161442.1 12, 13 1694
          NM_001167971.1 NP_001161443.1 12, 13 1675
          NM_001167972.1 NP_001161444.1 11, 12 1637
          NM_024917.5 NP_079193.2 12, 13 2087
          XM_005262195.2 XP_005262252.1 13, 14 1810
          XM_005262196.2 XP_005262253.1 11, 12 1605
          XM_005262201.2 XP_005262258.1 9, 10 1185
          XM_006724705.2 XP_006724768.1 12, 13 1749
          XM_011531042.2 XP_011529344.1 9, 10 1303
          XM_011531044.2 XP_011529346.1 11, 12 1364
          XM_017029858.1 XP_016885347.1 12, 13 2082
          XM_017029859.1 XP_016885348.1 11, 12 1397
          XM_017029860.1 XP_016885349.1 12, 13 1696
          XM_017029861.1 XP_016885350.1 11, 12 1606
          XM_017029862.1 XP_016885351.1 11, 12 1947
          XM_017029863.1 XP_016885352.1 13, 14 1619
          XM_017029864.1 XP_016885353.1 11, 12 1811
          XM_017029865.1 XP_016885354.1 11, 12 1439
          XM_017029866.1 XP_016885355.1 12, 13 1502
          XM_017029867.1 XP_016885356.1 12, 13 1539
          XM_017029868.1 XP_016885357.1 11, 12 1412
          XM_017029869.1 XP_016885358.1 10, 11 1756
          XM_017029870.1 XP_016885359.1 10, 11 1346
          XM_017029871.1 XP_016885360.1 10, 11 1328
          XM_017029872.1 XP_016885361.1 11, 12 1439
          XM_017029873.1 XP_016885362.1 11, 12 1414
          XM_017029874.1 XP_016885363.1 10, 11 1756
          XM_017029875.1 XP_016885364.1 9, 10 1303
          GenBank mRNA AK022749.1 - 11, 12 1450
          AK315780.1 - 12, 13 1844
          AL832849.1 - 12, 13 1613
          BC007526.1 - 11, 12 1593 2988022
          BC008067.2 - 5, 6 636
          BC009437.1 - 11, 12 1580 3504468
          BC020116.1 - 12, 13 1648 4342798
          BC034272.2 - 12, 13 2052 4933351
          BG747665.1 - 1, 2 73
          BX647969.1 - 11, 12 1634

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA methylation
          tRNA processing
          protein binding
          S-adenosylmethionine-dependent tRNA (m5U54) methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0