Search
Search

CAMK2A| Gene Symbol: CAMK2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015981.3 | NP_057065.2 | 16 | 1321 | ||
| NM_171825.2 | NP_741960.1 | 15 | 1288 | |||
| XM_017009898.1 | XP_016865387.1 | 15 | 1288 | |||
| GenBank mRNA | AB023185.1 | - | 15 | 1221 | ||
| AF091486.1 | - | 15 | 1177 | |||
| AF145710.1 | - | 15 | 1079 | |||
| AF145711.1 | - | 16 | 1112 | |||
| AK289776.1 | - | 16 | 1321 | |||
| BC040457.1 | - | 15 | 1203 | 4811948 | ||