Search
Search

FAM126A| Gene Symbol: FAM126A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032581.3 | NP_115970.2 | 4 | 477 | ||
| XM_005249894.3 | XP_005249951.1 | 4 | 521 | |||
| XM_005249895.2 | XP_005249952.1 | 4 | 521 | |||
| XM_011515589.2 | XP_011513891.1 | 4 | 328 | |||
| XM_011515590.1 | XP_011513892.1 | 4 | 521 | |||
| GenBank mRNA | AB030241.1 | - | 4 | 383 | ||
| AK056319.1 | - | 4 | 414 | |||
| AK297399.1 | - | 3 | 312 | |||
| AL833296.1 | - | 3 | 345 | |||
| BC018710.1 | - | 4 | 455 | 4649785 | ||
| DB099987.1 | - | 4 | 521 | |||
| KU178808.1 | - | 3 | 222 | |||
| KU178809.1 | - | 3 | 222 | |||
| KU178810.1 | - | 3 | 222 | |||