Search
Search

MCEE| Gene Symbol: MCEE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032601.3 | NP_115990.3 | 2 | 154 | ||
| XM_005264613.2 | XP_005264670.1 | 2 | 178 | |||
| GenBank mRNA | AF364547.1 | - | 2 | 127 | ||
| BC020825.1 | - | 2 | 119 | 4734422 | ||
| BM271757.1 | - | 2 | 176 | |||