Search
Search

COX14| Gene Symbol: COX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257133.1 | NP_001244062.1 | 3 | 653 | ||
| NM_001257134.1 | NP_001244063.1 | 2 | 789 | |||
| NM_032901.3 | NP_116290.1 | 2 | 593 | |||
| GenBank mRNA | AI208662.1 | - | 1 | 158 | ||
| BC007849.2 | - | 2 | 457 | 4135996 | ||
| BG829783.1 | - | 2 | 523 | |||