Search
Search

SIGLEC5| Gene Symbol: SIGLEC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003830.3 | NP_003821.1 | 6 | 1281 | ||
| XM_011527438.2 | XP_011525740.1 | 6 | 1235 | |||
| XM_017027419.1 | XP_016882908.1 | 6 | 1235 | |||
| GenBank mRNA | AF170484.1 | - | 6 | 1178 | ||
| AK225753.1 | - | 6 | 1202 | |||
| BC029896.1 | - | 6 | 1183 | 5178889 | ||
| U71383.1 | - | 6 | 1284 | |||