Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOCS2 SIRNA Assays ›
Gene Symbol:
SOCS2
Gene Name:
suppressor of cytokine signaling 2
Gene Aliases:
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2
Chromosome Location:
Chr.12: 93569822 - 93626236 on Build GRCh38
UniGene ID:
Hs.485572
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189602
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOCS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270467.1
NP_001257396.1
3
980
NM_001270468.1
NP_001257397.1
3
949
NM_001270469.1
NP_001257398.1
3
947
NM_001270470.1
NP_001257399.1
3
1219
NM_001270471.1
NP_001257400.1
2
872
NM_003877.4
NP_003868.1
3
1090
XM_011538929.1
XP_011537231.1
2
915
XM_011538935.1
XP_011537237.1
2
915
XM_011538936.1
XP_011537238.1
2
915
XM_017020147.1
XP_016875636.1
3
1063
XM_017020148.1
XP_016875637.1
3
953
XM_017020149.1
XP_016875638.1
3
947
XM_017020150.1
XP_016875639.1
3
1154
XM_017020151.1
XP_016875640.1
3
959
XM_017020152.1
XP_016875641.1
3
988
XM_017020153.1
XP_016875642.1
3
730
XM_017020154.1
XP_016875643.1
3
792
XM_017020155.1
XP_016875644.1
2
915
XM_017020156.1
XP_016875645.1
3
1205
GenBank mRNA
AB004903.1
-
2
538
AB006966.1
-
2
500
AB451318.1
-
2
500
AF020590.1
-
2
699
AF037989.1
-
3
817
AK290546.1
-
3
800
AK297662.1
-
4
680
AK313165.1
-
3
927
BC010399.1
-
2
712
4277425
BC070039.1
-
2
2605
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
negative regulation of protein kinase activity
JAK-STAT cascade
regulation of signal transduction
positive regulation of signal transduction
protein ubiquitination
cytokine-mediated signaling pathway
response to estradiol
cellular response to hormone stimulus
negative regulation of apoptotic process
negative regulation of JAK-STAT cascade
negative regulation of insulin receptor signaling pathway
growth hormone receptor signaling pathway
protein kinase inhibitor activity
SH3/SH2 adaptor activity
growth hormone receptor binding
insulin-like growth factor receptor binding
protein binding
JAK pathway signal transduction adaptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0