Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KALRN SIRNA Assays ›
          Gene Symbol:
          KALRN
          Gene Name:
          kalirin, RhoGEF kinase
          Gene Aliases:
          ARHGEF24, CHD5, CHDS5, DUET, DUO, HAPIP, TRAD
          Chromosome Location:
          Chr.3: 124033341 - 124726325 on Build GRCh38
          UniGene ID:
          Hs.13281
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS189736
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KALRN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024660.4 NP_001019831.2 9, 10 1803
          NM_001322988.1 NP_001309917.1 9, 10 1803
          NM_001322989.1 NP_001309918.1 9, 10 1803
          NM_001322990.1 NP_001309919.1 9, 10 1803
          NM_001322991.1 NP_001309920.1 9, 10 1803
          NM_001322992.1 NP_001309921.1 9, 10 1803
          NM_003947.5 NP_003938.1 9, 10 1803
          XM_006713810.3 XP_006713873.1 9, 10 2048
          XM_006713811.3 XP_006713874.1 9, 10 2048
          XM_006713812.3 XP_006713875.1 9, 10 2048
          XM_006713813.3 XP_006713876.1 9, 10 2048
          XM_006713814.3 XP_006713877.1 9, 10 2048
          XM_006713815.3 XP_006713878.1 9, 10 2051
          XM_011513279.2 XP_011511581.1 9, 10 2048
          XM_011513280.2 XP_011511582.1 9, 10 2049
          XM_011513281.2 XP_011511583.1 9, 10 24697
          XM_011513283.2 XP_011511585.1 9, 10 2045
          XM_011513285.2 XP_011511587.1 9, 10 2044
          XM_017007429.1 XP_016862918.1 9, 10 2048
          XM_017007430.1 XP_016862919.1 9, 10 2048
          XM_017007431.1 XP_016862920.1 9, 10 2044
          XM_017007432.1 XP_016862921.1 9, 10 2044
          XM_017007433.1 XP_016862922.1 9, 10 2051
          XM_017007434.1 XP_016862923.1 9, 10 2051
          XM_017007435.1 XP_016862924.1 9, 10 2051
          GenBank mRNA AK314126.1 - 9, 10 1803
          U94190.1 - 9, 10 1746

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          signal transduction
          nervous system development
          vesicle-mediated transport
          regulation of Rho protein signal transduction
          intracellular signal transduction
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          regulation of small GTPase mediated signal transduction
          protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0