Search
Search

TMEM55B| Gene Symbol: TMEM55B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100814.2 | NP_001094284.1 | 6 | 943 | ||
| NM_144568.3 | NP_653169.2 | 6 | 922 | |||
| GenBank mRNA | AK314021.1 | - | 6 | 923 | ||
| AL137727.1 | - | 2 | 2050 | |||
| BC002867.1 | - | 6 | 704 | 3940519 | ||
| BC020947.1 | - | 6 | 787 | 4816227 | ||
| BX161490.1 | - | 6 | 788 | |||
| BX248025.1 | - | 6 | 752 | |||