Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BOC SIRNA Assays ›
Gene Symbol:
BOC
Gene Name:
BOC cell adhesion associated, oncogene regulated
Gene Aliases:
CDON2
Chromosome Location:
Chr.3: 113211003 - 113287463 on Build GRCh38
UniGene ID:
Hs.591318
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189965
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BOC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301861.1
NP_001288790.1
12
2377
NM_033254.3
NP_150279.1
12
2725
XM_005247891.2
XP_005247948.1
12
2715
XM_005247892.2
XP_005247949.1
12
2711
XM_005247897.2
XP_005247954.1
11
2442
XM_011513305.2
XP_011511607.1
12
2712
XM_017007441.1
XP_016862930.1
12
2724
XM_017007442.1
XP_016862931.1
12
2720
XM_017007443.1
XP_016862932.1
12
2512
XM_017007444.1
XP_016862933.1
12
2304
XM_017007445.1
XP_016862934.1
12
2377
XM_017007446.1
XP_016862935.1
12
2344
XM_017007447.1
XP_016862936.1
12
2335
XM_017007448.1
XP_016862937.1
12
2256
XM_017007449.1
XP_016862938.1
12
2463
XM_017007450.1
XP_016862939.1
12
2721
XM_017007451.1
XP_016862940.1
12
2854
XM_017007452.1
XP_016862941.1
10
1887
XM_017007453.1
XP_016862942.1
10
1878
XM_017007454.1
XP_016862943.1
10
2263
GenBank mRNA
AK074556.1
-
5
1059
AK090455.1
-
6
4364
AY027658.1
-
11
2136
AY358328.1
-
13
2346
BC075829.1
-
12
2597
BC136390.1
-
11
2162
9020377
BX647184.1
-
6
2738
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
smoothened signaling pathway
axon guidance
positive regulation of myoblast differentiation
positive regulation of muscle cell differentiation
mitophagy in response to mitochondrial depolarization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0