Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBM39 SIRNA Assays ›
          Gene Symbol:
          RBM39
          Gene Name:
          RNA binding motif protein 39
          Gene Aliases:
          CAPER, CAPERalpha, FSAP59, HCC1, RNPC2
          Chromosome Location:
          Chr.20: 35703609 - 35742336 on Build GRCh38
          UniGene ID:
          Hs.282901
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS190435
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBM39
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242599.1 NP_001229528.1 5, 6 755
          NM_001242600.1 NP_001229529.1 5, 6 755
          NM_001323422.1 NP_001310351.1 6, 7 821
          NM_001323423.1 NP_001310352.1 8, 9 966
          NM_001323424.1 NP_001310353.1 6, 7 818
          NM_004902.3 NP_004893.1 6, 7 821
          NM_184234.2 NP_909122.1 6, 7 821
          NR_040722.1 7, 8 894
          NR_040723.1 7, 8 893
          NR_040724.1 7, 8 894
          XM_006723890.3 XP_006723953.1 6, 7 2401
          XM_006723891.3 XP_006723954.1 7, 8 1424
          XM_011529111.2 XP_011527413.1 7, 8 2473
          XM_017028138.1 XP_016883627.1 5, 6 755
          XM_017028139.1 XP_016883628.1 8, 9 1790
          XM_017028140.1 XP_016883629.1 6, 7 828
          XM_017028141.1 XP_016883630.1 6, 7 2401
          XM_017028142.1 XP_016883631.1 7, 8 3297
          XM_017028143.1 XP_016883632.1 7, 8 2473
          XM_017028144.1 XP_016883633.1 8, 9 966
          XM_017028145.1 XP_016883634.1 6, 7 828
          XM_017028146.1 XP_016883635.1 7, 8 894
          XM_017028147.1 XP_016883636.1 7, 8 817
          GenBank mRNA AK126158.1 - 3, 4 980
          AK297520.1 - 1, 2 192
          AK299165.1 - 5, 6 655
          AK299678.1 - 5, 6 657
          AK303910.1 - 6, 7 693
          AL833168.1 - 6, 7 707
          BC107886.1 - 7, 8 603
          BC131543.1 - 5, 6 521 40085367
          BC141835.1 - 6, 7 671 40148871
          BC158172.1 - 6, 7 671 40148190
          BX537770.1 - 7, 8 813
          BX640714.1 - 7, 8 806
          BX640812.1 - 7, 8 803
          CR749443.1 - 6, 7 2488
          L10910.1 - 6, 7 561
          L10911.1 - 5, 6 436

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          RNA processing
          mRNA processing
          RNA splicing
          nucleotide binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0