Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP6R2 SIRNA Assays ›
          Gene Symbol:
          PPP6R2
          Gene Name:
          protein phosphatase 6 regulatory subunit 2
          Gene Aliases:
          KIAA0685, PP6R2, SAP190, SAPS2
          Chromosome Location:
          Chr.22: 50343317 - 50445090 on Build GRCh38
          UniGene ID:
          Hs.449098
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS190542
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP6R2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242898.1 NP_001229827.1 17 2183
          NM_001242899.1 NP_001229828.1 16 2105
          NM_001242900.1 NP_001229829.1 16 2102
          NM_014678.4 NP_055493.2 16 2102
          XM_005261960.3 XP_005262017.1 11 1137
          XM_006724431.2 XP_006724494.1 18 2294
          XM_006724434.1 XP_006724497.1 19 2443
          XM_006724435.2 XP_006724498.1 16 2087
          XM_011530720.2 XP_011529022.1 16 1871
          XM_011530721.2 XP_011529023.1 16 1868
          XM_011530722.2 XP_011529024.1 16 1868
          XM_011530723.2 XP_011529025.1 16 1871
          XM_011530724.2 XP_011529026.1 16 1871
          XM_011530726.2 XP_011529028.1 15 1790
          XM_011530727.2 XP_011529029.1 15 1790
          XM_011530728.2 XP_011529030.1 15 1790
          XM_011530729.2 XP_011529031.1 15 1790
          XM_011530730.2 XP_011529032.1 15 1787
          XM_011530731.1 XP_011529033.1 15 1787
          XM_011530732.2 XP_011529034.1 19 2347
          XM_011530734.2 XP_011529036.1 18 2221
          XM_011530735.2 XP_011529037.1 17 2171
          XM_011530736.2 XP_011529038.1 18 2297
          XM_011530737.2 XP_011529039.1 19 2446
          XM_011530738.2 XP_011529040.1 18 2249
          XM_011530739.2 XP_011529041.1 16 1911
          XM_011530740.2 XP_011529042.1 18 2319
          XM_017029116.1 XP_016884605.1 15 1787
          XM_017029117.1 XP_016884606.1 20 2496
          XM_017029118.1 XP_016884607.1 20 2448
          XM_017029119.1 XP_016884608.1 19 2370
          XM_017029120.1 XP_016884609.1 19 2344
          XM_017029121.1 XP_016884610.1 17 2168
          XM_017029122.1 XP_016884611.1 18 2246
          XM_017029123.1 XP_016884612.1 17 2168
          XM_017029124.1 XP_016884613.1 18 2294
          XM_017029125.1 XP_016884614.1 17 2168
          XM_017029126.1 XP_016884615.1 17 2171
          XM_017029127.1 XP_016884616.1 18 2246
          XM_017029128.1 XP_016884617.1 18 2294
          XM_017029129.1 XP_016884618.1 16 2087
          XM_017029130.1 XP_016884619.1 16 2087
          XM_017029131.1 XP_016884620.1 17 2213
          XM_017029132.1 XP_016884621.1 17 2165
          XM_017029133.1 XP_016884622.1 17 2125
          XM_017029134.1 XP_016884623.1 17 2122
          XM_017029135.1 XP_016884624.1 18 2172
          XM_017029136.1 XP_016884625.1 17 2122
          XM_017029137.1 XP_016884626.1 17 2122
          XM_017029138.1 XP_016884627.1 16 2041
          GenBank mRNA AB014585.1 - 16 2074
          AK002063.1 - 10 979
          AK022669.1 - 6 606
          AK302472.1 - 17 2094
          BC000976.4 - 18 2172 2900437
          BC006568.2 - 16 2088 3532211
          BC032664.1 - 16 2102 5553142
          BC041698.1 - 16 2072 5732101
          BC052995.2 - 16 2008 6162781

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0