Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS190581
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
12, 13
1824
NM_001204145.2
NP_001191074.1
10, 11
1516
NM_001204146.2
NP_001191075.1
10, 11
1694
NM_001204147.2
NP_001191076.1
9, 10
1476
NM_001204148.2
NP_001191077.1
10, 11
1623
NM_001321868.1
NP_001308797.1
11, 12
1764
NM_001321869.1
NP_001308798.1
12, 13
1905
NM_001321870.1
NP_001308799.1
12, 13
1896
NM_001321871.1
NP_001308800.1
11, 12
1779
NM_001321872.1
NP_001308801.1
11, 12
1773
NM_001321873.1
NP_001308802.1
11, 12
1767
NM_001321874.1
NP_001308803.1
11, 12
1826
NM_001321875.1
NP_001308804.1
11, 12
1832
NM_001321877.1
NP_001308806.1
10, 11
1694
NM_001321879.1
NP_001308808.1
10, 11
1688
NM_001321884.1
NP_001308813.1
11, 12
1820
NM_001321885.1
NP_001308814.1
10, 11
1709
NM_001321886.1
NP_001308815.1
12, 13
1876
NM_001321887.1
NP_001308816.1
12, 13
1710
NM_001321888.1
NP_001308817.1
10, 11
1510
NM_001321889.1
NP_001308818.1
11, 12
1747
NM_001321890.1
NP_001308819.1
11, 12
1811
NM_001321891.1
NP_001308820.1
10, 11
1685
NM_001321893.1
NP_001308822.1
11, 12
1582
NM_001321894.1
NP_001308823.1
11, 12
1591
NM_001321895.1
NP_001308824.1
10, 11
1525
NM_001321896.1
NP_001308825.1
12, 13
1995
NM_001321897.1
NP_001308826.1
11, 12
1591
NM_001321898.1
NP_001308827.1
10, 11
1540
NM_001321899.1
NP_001308828.1
12, 13
1723
NM_001321900.1
NP_001308829.1
11, 12
1817
NM_001321901.1
NP_001308830.1
11, 12
1651
NM_001321902.1
NP_001308831.1
11, 12
1657
NM_014707.3
NP_055522.1
11, 12
1648
NM_058176.2
NP_478056.1
10, 11
1503
NM_178423.2
NP_848510.1
11, 12
1648
NM_178425.3
NP_848512.1
10, 11
1512
XM_011515629.2
XP_011513931.1
11, 12
1606
XM_011515631.2
XP_011513933.1
12, 13
1920
XM_011515634.2
XP_011513936.1
10, 11
1474
XM_011515635.2
XP_011513937.1
12, 13
2472
XM_011515636.2
XP_011513938.1
11, 12
2193
XM_011515637.2
XP_011513939.1
11, 12
1617
XM_011515638.2
XP_011513940.1
11, 12
2187
XM_011515639.2
XP_011513941.1
11, 12
2384
XM_011515641.2
XP_011513943.1
12, 13
2469
XM_011515642.2
XP_011513944.1
10, 11
3198
XM_011515644.2
XP_011513946.1
12, 13
2444
XM_011515645.2
XP_011513947.1
10, 11
3183
XM_011515646.2
XP_011513948.1
10, 11
3174
XM_011515648.2
XP_011513950.1
11, 12
2202
XM_011515649.2
XP_011513951.1
11, 12
2406
XM_011515651.1
XP_011513953.1
10, 11
1440
XM_017012826.1
XP_016868315.1
11, 12
2178
XM_017012827.1
XP_016868316.1
12, 13
2261
XM_017012828.1
XP_016868317.1
10, 11
2254
XM_017012829.1
XP_016868318.1
11, 12
1581
XM_017012830.1
XP_016868319.1
11, 12
1572
XM_017012831.1
XP_016868320.1
10, 11
1449
GenBank mRNA
AB018287.1
-
11, 12
1612
AJ459808.1
-
11, 12
1612
AK097178.1
-
10, 11
1474
AK296214.1
-
10, 11
1426
AK297404.1
-
10, 11
1470
AK304298.1
-
12, 13
1817
AK304343.1
-
10, 11
1543
AK304410.1
-
9, 10
1439
AK316109.1
-
10, 11
1645
AY032737.1
-
10, 11
1462
AY032738.1
-
10, 11
1462
AY197371.1
-
10, 11
1471
BC111735.1
-
9, 10
1338
BC150328.1
-
11, 12
1623
8860495
BC152405.1
-
11, 12
1612
8862508
CR749550.1
-
10, 11
1579
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0